5-Fluoro-2-formylpyridine
Supplier: Apollo Scientific
Warning
Synonyms: 5-Fluoropyridine-2-carboxaldehyde, 5-Fluoropicolinaldehyde
PC7042-1G PC7042-5G PC7042-10G
APOSPC7042-1GEA 60.9 EUR
APOSPC7042-1G APOSPC7042-5G APOSPC7042-10G
5-Fluoro-2-formylpyridine
5-Fluoro-2-formylpyridine
Formula: C₆H₄FNO
MW: 125.1 g/mol
Boiling Pt: 72…74 °C/21mmHg
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD07781242
CAS Number: 31181-88-1