2-Fluoro-3-methylphenylboronic acid
Supplier: Apollo Scientific
Warning
Synonyms: (2-Fluoro-3-methylphenyl)boronic acid
PC7045-1G PC7045-5G PC7045-25G
APOSPC7045-1GEA 21.9 EUR
APOSPC7045-1G APOSPC7045-5G APOSPC7045-25G
2-Fluoro-3-methylphenylboronic acid
2-Fluoro-3-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
Melting Pt: 77…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06409950
CAS Number: 762287-58-1