Potassium-2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
Supplier: Apollo Scientific
Synonyms: Potassium 2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
PC7058-1G PC7058-5G
APOSPC7058-1GEA 94.5 EUR
APOSPC7058-1G APOSPC7058-5G
Potassium-2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
Potassium 2-fluoro-4-(trifluoromethyl)phenyltrifluoroborate
Formula: C₇H₃BF₇K
MW: 269.9969 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781236
CAS Number: 1150655-12-1