N-Methyl-N-[4-(trifluoromethyl)phenyl]amine 98%
Supplier: Apollo Scientific
Warning
Synonyms: N-Methyl-4-(trifluoromethyl)aniline
PC7074-1G PC7074-5G
APOSPC7074-1GEA 21.9 EUR
APOSPC7074-1G APOSPC7074-5G
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine 98%
N-Methyl-N-[4-(trifluoromethyl)phenyl]amine
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 105…106 °C/20mmHg
Density: 1.2394 g/cm³ (25 °C)
Flash Pt: 99 °C
Storage Temperature: Ambient
MDL Number: MFCD07369717
CAS Number: 22864-65-9