2-Ethoxy-4-fluoro-1-nitrobenzene 98%
Supplier: Apollo Scientific
Warning
PC7082-1G PC7082-5G
APOSPC7082-1GEA 21.9 EUR
APOSPC7082-1G APOSPC7082-5G
2-Ethoxy-4-fluoro-1-nitrobenzene 98%
2-Ethoxy-4-fluoro-1-nitrobenzene
Formula: C₈H₈FNO₃
MW: 185.1524 g/mol
Melting Pt: 35…37 °C
Storage Temperature: Ambient
MDL Number: MFCD04115630
CAS Number: 28987-44-2