(3-Fluorophenyl)methanesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
PC7103-1G PC7103-250MG
APOSPC7103-1GEA 87 EUR
APOSPC7103-1G APOSPC7103-250MG
(3-Fluorophenyl)methanesulphonyl chloride 97%
(3-Fluorophenyl)methanesulphonyl chloride
Formula: C₇H₆ClFO₂S
Melting Pt: 39…41 °C
Storage Temperature: Ambient
MDL Number: MFCD04117456
CAS Number: 24974-72-9
UN: 3261
ADR: 8,II