α-Chloro-2,4,5-trifluorotoulene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4,5-Trifluorobenzyl chloride, 1-(Chloromethyl)-2,4,5-trifluorobenzene
PC7113-100G PC7113-25G PC7113-500G
APOSPC7113-100GEA 435 EUR
APOSPC7113-100G APOSPC7113-25G APOSPC7113-500G
α-Chloro-2,4,5-trifluorotoulene 98%
α-Chloro-2,4,5-trifluorotoulene
Formula: C₇H₄ClF₃
MW: 180.56 g/mol
Density: 1.689 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00236319
CAS Number: 243139-71-1
UN: 3265
ADR: 8 II