4-Benzyloxy-2-fluoro-1-nitrobenzene 98%
Supplier: Apollo Scientific
Synonyms: 4-(Benzyloxy)-2-fluoro-1-nitrobenzene
PC7128-1G PC7128-5G
APOSPC7128-1GEA 198 EUR
APOSPC7128-1G APOSPC7128-5G
4-Benzyloxy-2-fluoro-1-nitrobenzene 98%
4-Benzyloxy-2-fluoro-1-nitrobenzene
Formula: C₁₃H₁₀FNO₃
MW: 247.2200 g/mol
Melting Pt: 85…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115634
CAS Number: 221040-07-9