2,3-Dichloro-4-fluoroaniline 95%
Supplier: Apollo Scientific
|
Warning
|
PC7141-25G
PC7141-5G
APOSPC7141-25GEA
488
EUR
APOSPC7141-25G
APOSPC7141-5G
2,3-Dichloro-4-fluoroaniline 95%
2,3-Dichloro-4-fluoroaniline
Formula:
C₆H₄Cl₂FN MW: 180.0071 g/mol Melting Pt: 63…68 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08706409 CAS Number: 36556-52-2 |