Trifluoroacetamide 98+%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,2-Trifluoroacetamide
PC7150-100G PC7150-500G PC7150-1KG
APOSPC7150-100GEA 21.9 EUR
APOSPC7150-100G APOSPC7150-500G APOSPC7150-1KG
Trifluoroacetamide 98+%
Trifluoroacetamide
Formula: C₂H₂F₃NO
MW: 113.04 g/mol
Boiling Pt: 162…164 °C
Melting Pt: 65…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00008008
CAS Number: 354-38-1
EINECS: 206-559-9