Trifluoroacetic acid 99,5%
Supplier: Apollo Scientific
Danger
Synonyms: TFA
PC7160A-1L PC7160A-500ML
APOSPC7160A-1LEA 92.8 EUR
APOSPC7160A-1L APOSPC7160A-500ML
Trifluoroacetic acid 99,5%
Trifluoroacetic acid
Plastic coated bottle with tamper evident cap
Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 72.4 °C
Melting Pt: –15.4 °C
Density: 1.48 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8 I
Merck Index: 13,09754