4-Fluorophenethyl bromide 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Fluoro-4-(2-bromoethyl)benzene, 1-(2-bromoethyl)-4-fluorobenzene
PC7166-25G PC7166-5G PC7166-10G
APOSPC7166-25GEA 58 EUR
APOSPC7166-25G APOSPC7166-5G APOSPC7166-10G
4-Fluorophenethyl bromide 97%
4-Fluorophenethyl bromide
Formula: C₈H₈BrF
MW: 203.05 g/mol
Boiling Pt: 100…104 °C/15mmHg
Density: 1.4498 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD03844794
CAS Number: 332-42-3