5-Amino-3-fluorobenzonitrile 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-Amino-5-fluorobenzonitrile, 3-Cyano-5-fluoroaniline
PC7168-5G PC7168-25G
APOSPC7168-5GEA 55.1 EUR
APOSPC7168-5G APOSPC7168-25G
5-Amino-3-fluorobenzonitrile 97%
5-Amino-3-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Melting Pt: 105…107 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD05864357
CAS Number: 210992-28-2
UN: 3439
ADR: 6.1 III