1-(5-Bromo-2,4-difluorophenyl)ethanone 98%
Supplier: Apollo Scientific
Warning
PC7171-1G PC7171-25G PC7171-5G
APOSPC7171-1GEA 21.9 EUR
APOSPC7171-1G APOSPC7171-25G APOSPC7171-5G
1-(5-Bromo-2,4-difluorophenyl)ethanone 98%
1-(5-Bromo-2,4-difluorophenyl)ethanone
Formula: C₈H₅BrF₂O
Melting Pt: 31…32 °C
Storage Temperature: Ambient
MDL Number: MFCD13190613
CAS Number: 864773-64-8