2,4-Difluoro-5-nitrophenylboronic acid 97%
Supplier: Apollo Scientific
Warning
PC7184-1G PC7184-250MG
APOSPC7184-1GEA 306 EUR
APOSPC7184-1G APOSPC7184-250MG
2,4-Difluoro-5-nitrophenylboronic acid 97%
2,4-Difluoro-5-nitrophenylboronic acid
Formula: C₆H₄BF₂NO₄
MW: 202.91 g/mol
Melting Pt: 225…227 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03095253
CAS Number: 325786-11-6