1,1,1-Trifluoroacetylacetone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1-Trifluoropentane-2,4-dione, 1,1,1-Trifluoro-2,4-pentanedione
PC7210-100G PC7210-5G
APOSPC7210-100GEA 94.4 EUR
APOSPC7210-100G APOSPC7210-5G
1,1,1-Trifluoroacetylacetone 98%
1,1,1-Trifluoroacetylacetone
Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 107 °C
Density: 1.27 g/cm³ (25 °C)
Flash Pt: 26 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000427
CAS Number: 367-57-7
EINECS: 206-698-5
UN: 1224
ADR: 3,III