4-(Trifluoroacetyl)benzoic acid 97+%
Supplier: Apollo Scientific
Warning
Synonyms: 4-(2,2,2-trifluoroacetyl)benzoic acid
PC7214-1G PC7214-25G PC7214-5G
APOSPC7214-1GEA 50.9 EUR
APOSPC7214-1G APOSPC7214-25G APOSPC7214-5G
4-(Trifluoroacetyl)benzoic acid 97+%
4-(Trifluoroacetyl)benzoic acid
Formula: C₉H₅F₃O₃
MW: 218.13 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052340
CAS Number: 58808-59-6