1,3-Dimethyl-8-(trifluoromethyl)-1,5-dihydropyrido[3,2-d]pyrimidine-2,4,6(3H)-trione
Supplier: Apollo Scientific
Warning
PC7217-10G PC7217-1G PC7217-5G
APOSPC7217-10GEA 1350 EUR
APOSPC7217-10G APOSPC7217-1G APOSPC7217-5G
1,3-Dimethyl-8-(trifluoromethyl)-1,5-dihydropyrido[3,2-d]pyrimidine-2,4,6(3H)-trione
1,3-Dimethyl-8-(trifluoromethyl)-1,5-dihydropyrido[3,2-d]pyrimidine-2,4,6(3H)-trione
Formula: C₁₀H₈F₃N₃O₃
MW: 275.19 g/mol
Melting Pt: 171…174 °C
Storage Temperature: Ambient
MDL Number: MFCD09607925
CAS Number: 952183-38-9