1,3-Dimethyl-4-(trifluoromethyl)-1,7-dihydro-6H-pyrazolo[3,4-b]pyridin-6-one
Supplier: Apollo Scientific
PC7219-10G PC7219-1G PC7219-5G
APOSPC7219-10GEA 1350 EUR
APOSPC7219-10G APOSPC7219-1G APOSPC7219-5G
1,3-Dimethyl-4-(trifluoromethyl)-1,7-dihydro-6H-pyrazolo[3,4-b]pyridin-6-one
1,3-Dimethyl-4-(trifluoromethyl)-1,7-dihydro-6H-pyrazolo[3,4-b]pyridin-6-one
Formula: C₉H₈F₃N₃O
MW: 231.18 g/mol
Melting Pt: 235…237 °C
Storage Temperature: Ambient
MDL Number: MFCD06195730
CAS Number: 94835-66-2