3,4-Difluorobenzenesulphonamide
Supplier: Apollo Scientific
Warning
PC7245-1G PC7245-5G PC7245-25G
APOSPC7245-1GEA 21.9 EUR
APOSPC7245-1G APOSPC7245-5G APOSPC7245-25G
3,4-Difluorobenzenesulphonamide
3,4-Difluorobenzenesulphonamide
Formula: C₆H₅F₂NO₂S
MW: 193.17 g/mol
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00728800
CAS Number: 108966-71-8