2,3,4-Trifluoroaniline 99%
Supplier: Apollo Scientific
Warning
PC7258-100G PC7258-250G PC7258-25G PC7258-500G
APOSPC7258-100GEA 24.8 EUR
APOSPC7258-100G APOSPC7258-250G APOSPC7258-25G APOSPC7258-500G
2,3,4-Trifluoroaniline 99%
2,3,4-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 90…92 °C/48mmHg
Density: 1.393 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00011737
CAS Number: 3862-73-5
EINECS: 407-170-9
UN: 3082
ADR: 9+EHS III