2,3,6-Trifluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC7258L-1G PC7258L-5G PC7258L-25G
APOSPC7258L-1GEA 26.1 EUR
APOSPC7258L-1G APOSPC7258L-5G APOSPC7258L-25G
2,3,6-Trifluoroaniline 98%
2,3,6-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 150…151 °C
Density: 1.39 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00012369
CAS Number: 67815-56-9