4-Bromo-2-fluoropyridine
Supplier: Apollo Scientific
Danger
PC7262-1G PC7262-25G PC7262-5G PC7262-100G
APOSPC7262-1GEA 0 EUR
APOSPC7262-1G APOSPC7262-25G APOSPC7262-5G APOSPC7262-100G
4-Bromo-2-fluoropyridine
4-Bromo-2-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
Boiling Pt: 65 °C/5mmHg
Density: 1.713 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD04112504
CAS Number: 128071-98-7