2,5-Dichloro-4-(trifluoromethyl)pyridine
Supplier: Apollo Scientific
Danger
PC7264-1G PC7264-500MG
APOSPC7264-1GEA 386 EUR
APOSPC7264-1G APOSPC7264-500MG
2,5-Dichloro-4-(trifluoromethyl)pyridine
2,5-Dichloro-4-(trifluoromethyl)pyridine
Formula: C₆H₂Cl₂F₃N
MW: 215.99 g/mol
Boiling Pt: 76 °C
Density: 1.542 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD09749663
CAS Number: 89719-92-6
UN: 2810
ADR: 6.1,III