2,3,4-Trifluorobenzaldehyde 97%
Supplier: Apollo Scientific
Danger
PC7265D-100G PC7265D-25G PC7265D-5G
APOSPC7265D-100GEA 206 EUR
APOSPC7265D-100G APOSPC7265D-25G APOSPC7265D-5G
2,3,4-Trifluorobenzaldehyde 97%
2,3,4-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 77…79 °C/20mmHg
Density: 1.404 g/cm³ (25 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00061230
CAS Number: 161793-17-5
UN: 1989
ADR: 3 III