2,4,5-Trifluorobenzaldehyde 98%
Supplier: Apollo Scientific
Warning
PC7265J-100G PC7265J-50G
APOSPC7265J-100GEA 120 EUR
APOSPC7265J-100G APOSPC7265J-50G
2,4,5-Trifluorobenzaldehyde 98%
2,4,5-Trifluorobenzaldehyde
Formula: C₇H₃F₃O
MW: 160.1 g/mol
Boiling Pt: 174 °C
Density: 1.408 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00061196
CAS Number: 165047-24-5
UN: 1989
ADR: 3,III