2,4,5-Trifluorobenzamide 97%
Supplier: Apollo Scientific
Warning
PC7265W-25G PC7265W-5G PC7265W-1G
APOSPC7265W-25GEA 241 EUR
APOSPC7265W-25G APOSPC7265W-5G APOSPC7265W-1G
2,4,5-Trifluorobenzamide 97%
2,4,5-Trifluorobenzamide
Formula: C₇H₄F₃NO
MW: 175.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082478
CAS Number: 98349-23-6