1,2,3-Trifluorobenzene
Supplier: Apollo Scientific
Danger
PC7269-100G PC7269-25G
APOSPC7269-100GEA 78.3 EUR
APOSPC7269-100G APOSPC7269-25G
1,2,3-Trifluorobenzene
1,2,3-Trifluorobenzene
Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 94…95 °C
Density: 1.28 g/cm³ (25 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00012013
CAS Number: 1489-53-8
UN: 1993
ADR: 3,II