1,2,4-Trifluorobenzene 99%
Supplier: Apollo Scientific
Danger
PC7270-100G PC7270-25G PC7270-500G
APOSPC7270-100GEA 40.6 EUR
APOSPC7270-100G APOSPC7270-25G APOSPC7270-500G
1,2,4-Trifluorobenzene 99%
1,2,4-Trifluorobenzene
Formula: C₆H₃F₃
MW: 132.09 g/mol
Boiling Pt: 89…91 °C
Melting Pt: -35
Density: 1.264 g/cm³ (25 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000305
CAS Number: 367-23-7
EINECS: 206-684-9
UN: 1993
ADR: 3 II