1,3,5-Trifluorobenzene 98%
Supplier: Apollo Scientific
Danger
PC7280-100G PC7280-25G PC7280-5G
APOSPC7280-100GEA 130 EUR
APOSPC7280-100G APOSPC7280-25G APOSPC7280-5G
1,3,5-Trifluorobenzene 98%
1,3,5-Trifluorobenzene
Formula: C₆H₃F₃
Boiling Pt: 76 °C
Melting Pt: –5.5 °C
Density: 1.277 g/cm³ (25 °C)
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00000333
CAS Number: 372-38-3
EINECS: 206-751-2
UN: 2387
ADR: 3,II