2,3,4-Trifluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2,3,4-Trifluorobenzene-1-sulphonyl chloride
PC7281-25G PC7281-5G PC7281-10G
APOSPC7281-25GEA 59.6 EUR
APOSPC7281-25G APOSPC7281-5G APOSPC7281-10G
2,3,4-Trifluorobenzenesulphonyl chloride 97%
Trifluorobenzenesulphonyl chloride
Formula: C₆H₂ClF₃O₂S
MW: 230.59 g/mol
Boiling Pt: 234…246 °C
Density: 1.64 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00102573
CAS Number: 175278-08-7
UN: 3265
ADR: 8,II