2,4,5-Trifluorobenzonitrile 99%
Supplier: Apollo Scientific
Warning
PC7283M-100G PC7283M-25G PC7283M-5G PC7283M-500G
APOSPC7283M-100GEA 183 EUR
APOSPC7283M-100G APOSPC7283M-25G APOSPC7283M-5G APOSPC7283M-500G
2,4,5-Trifluorobenzonitrile 99%
2,4,5-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 168…170 °C
Density: 1.373 g/cm³ (25 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00013289
CAS Number: 98349-22-5
UN: 3276
ADR: 6.1 III