2,4,6-Trifluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC7283R-100G PC7283R-25G PC7283R-5G
APOSPC7283R-100GEA 155 EUR
APOSPC7283R-100G APOSPC7283R-25G APOSPC7283R-5G
2,4,6-Trifluorobenzonitrile 98%
2,4,6-Trifluorobenzonitrile
Formula: C₇H₂F₃N
Boiling Pt: 179 °C
Melting Pt: 55…63 °C
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00042399
CAS Number: 96606-37-0
UN: 3439
ADR: 6.1,III