3,4,5-Trifluorobenzonitrile 97%
Supplier: Apollo Scientific
Warning
PC7283T-25G PC7283T-100G
APOSPC7283T-25GEA 28.7 EUR
APOSPC7283T-25G APOSPC7283T-100G
3,4,5-Trifluorobenzonitrile 97%
3,4,5-Trifluorobenzonitrile
Formula: C₇H₂F₃N
Boiling Pt: 72…73 °C/10mmHg
Melting Pt: 45…47 °C
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00074948
CAS Number: 134227-45-5
UN: 3439
ADR: 6.1,III