2,3,4-Trifluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,3,4-trifluorobenzoylchloride
PC7285-100G PC7285-25G PC7285-5G
APOSPC7285-100GEA 429 EUR
APOSPC7285-100G APOSPC7285-25G APOSPC7285-5G
2,3,4-Trifluorobenzoyl chloride 98%
2,3,4-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Boiling Pt: 143…144 °C/130mmHg
Density: 1.522 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00075078
CAS Number: 157373-08-5
UN: 3265
ADR: 8 II