2,3,6-Trifluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Danger
PC7285C-25G PC7285C-50G PC7285C-5G
APOSPC7285C-25GEA 284 EUR
APOSPC7285C-25G APOSPC7285C-50G APOSPC7285C-5G
2,3,6-Trifluorobenzoyl chloride 98%
2,3,6-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
Boiling Pt: 180…182 °C
Density: 1.48 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00061203
CAS Number: 189807-20-3
UN: 3265
ADR: 8,II