2,4,5-Trifluorobenzoyl chloride 99%
Supplier: Apollo Scientific
Danger
Synonyms: 2,4,5-trifluorobenzoylchloride
PC7285G-100G PC7285G-25G PC7285G-5G
APOSPC7285G-100GEA 186 EUR
APOSPC7285G-100G APOSPC7285G-25G APOSPC7285G-5G
2,4,5-Trifluorobenzoyl chloride 99%
2,4,5-Trifluorobenzoyl chloride
Formula: C₇H₂ClF₃O
MW: 194.54 g/mol
Boiling Pt: 85 °C/17mmHg
Density: 1.52 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00061204
CAS Number: 88419-56-1
UN: 3265
ADR: 8 II