2,3,6-Trifluorobenzyl alcohol 97%
Supplier: Apollo Scientific
Warning
PC7285LB-5G PC7285LB-100G PC7285LB-25G
APOSPC7285LB-5GEA 29 EUR
APOSPC7285LB-5G APOSPC7285LB-100G APOSPC7285LB-25G
2,3,6-Trifluorobenzyl alcohol 97%
2,3,6-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
Melting Pt: 43…44 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061205
CAS Number: 114152-19-1