3,4,5-Trifluorobenzyl alcohol 97%
Supplier: Apollo Scientific
Warning
PC7285LH-1G PC7285LH-5G
APOSPC7285LH-1GEA 21.9 EUR
APOSPC7285LH-1G APOSPC7285LH-5G
3,4,5-Trifluorobenzyl alcohol 97%
3,4,5-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 203…204 °C
Density: 1.385 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00083527
CAS Number: 220227-37-2