2,4,5-Trifluorobenzyl bromide 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-(Bromomethyl)-2,4,5-trifluorobenzene, α-Bromo-2,4,5-trifluorotoluene
PC7285S-100G PC7285S-25G PC7285S-500G PC7285S-1KG
APOSPC7285S-100GEA 203 EUR
APOSPC7285S-100G APOSPC7285S-25G APOSPC7285S-500G APOSPC7285S-1KG
2,4,5-Trifluorobenzyl bromide 97%
2,4,5-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
MW: 225.01 g/mol
Boiling Pt: 182
Melting Pt: 21
Density: 1.71 g/cm³ (25 °C)
Flash Pt: 90.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00061209
CAS Number: 157911-56-3
UN: 3265
ADR: 8 II