3,4,5-Trifluorobenzyl bromide 98%
Supplier: Apollo Scientific
Danger
Synonyms: 5-(Bromomethyl)-1,2,3-trifluorobenzene, α-Bromo-3,4,5-trifluorotoluene
PC7285V-25G PC7285V-5G PC7285V-100G
APOSPC7285V-25GEA 95.7 EUR
APOSPC7285V-25G APOSPC7285V-5G APOSPC7285V-100G
3,4,5-Trifluorobenzyl bromide 98%
3,4,5-Trifluorobenzyl bromide
Formula: C₇H₄BrF₃
Boiling Pt: 44…46 °C/1mmHg
Density: 1.689 g/cm³ (25 °C)
Flash Pt: 65.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00083528
CAS Number: 220141-72-0
UN: 3265
ADR: 8,II