4,4,4-Trifluoro-1-butanol 97%
Supplier: Apollo Scientific
Warning
PC7289-25G PC7289-5G PC7289-1G
APOSPC7289-25GEA 290 EUR
APOSPC7289-25G APOSPC7289-5G APOSPC7289-1G
4,4,4-Trifluoro-1-butanol 97%
4,4,4-Trifluoro-1-butanol
Formula: C₄H₇F₃O
Boiling Pt: 122…125 °C
Density: 1.206 g/cm³ (25 °C)
Flash Pt: 52 °C
Storage Temperature: Ambient
MDL Number: MFCD00041483
CAS Number: 461-18-7
UN: 1987
ADR: 3,III