1,1,1-Trifluoro-2-butanone 95%
Supplier: Apollo Scientific
Warning
PC7289T-100G PC7289T-25G
APOSPC7289T-100GEA 496 EUR
APOSPC7289T-100G APOSPC7289T-25G
1,1,1-Trifluoro-2-butanone 95%
1,1,1-Trifluoro-2-butanone
Formula: C₄H₅F₃O
MW: 126.08 g/mol
Boiling Pt: 44…45 °C
Density: 0.929 g/cm³ (25 °C)
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00190641
CAS Number: 381-88-4
UN: 1224
ADR: 3,II