4,4,4-Trifluorobut-2-en-1-ol 95%
Supplier: Apollo Scientific
Danger
Synonyms: 4,4,4-Trifluorocrotyl alcohol
PC7291L-5G
APOSPC7291L-5GEA 168 EUR
APOSPC7291L-5G
4,4,4-Trifluorobut-2-en-1-ol 95%
4,4,4-Trifluorobut-2-en-1-ol
Formula: C₄H₅F₃O
MW: 126.08 g/mol
Boiling Pt: 118.5 °C/630mmHg
Flash Pt: 37 °C
Storage Temperature: Ambient
MDL Number: MFCD00077603
CAS Number: 674-53-3
EINECS: 614-064-8
UN: 1987
ADR: 3,III