4,4,4-Trifluorocrotononitrile 97%
Supplier: Apollo Scientific
|
Danger
|
PC7293N-25G
PC7293N-5G
APOSPC7293N-25GEA
717
EUR
APOSPC7293N-25G
APOSPC7293N-5G
4,4,4-Trifluorocrotononitrile 97%
4,4,4-Trifluorocrotononitrile
Formula:
CF₃CH=CHCN MW: 121.06 g/mol Boiling Pt: 84…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190645 CAS Number: 406-86-0 UN: 3276 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...