1,1,1-Trifluoro-2,3-epoxypropane 98%
Supplier: Apollo Scientific
Danger
Synonyms: 3,3,3-Trifluoro-1,2-propenoxide
PC7296-25G PC7296-5G PC7296-1G PC7296-100G
APOSPC7296-25GEA 203 EUR
APOSPC7296-25G APOSPC7296-5G APOSPC7296-1G APOSPC7296-100G
1,1,1-Trifluoro-2,3-epoxypropane 98%
1,1,1-Trifluoro-2,3-epoxypropane
Formula: C₃H₃F₃O
MW: 112.05 g/mol
Boiling Pt: 39.1…39.3 °C/748mmHg
Melting Pt: –80 °C
Density: 1.33 g/cm³ (25 °C)
Flash Pt: –25 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041506
CAS Number: 359-41-1
UN: 1992
ADR: 3+6.1,II