4-Fluoro-2-nitrobenzonitrile
Supplier: Apollo Scientific
PC7339-1G PC7339-250MG
APOSPC7339-1GEA 29.3 EUR
APOSPC7339-1G APOSPC7339-250MG
4-Fluoro-2-nitrobenzonitrile
4-Fluoro-2-nitrobenzonitrile
Formula: C₇H₃FN₂O₂
MW: 166.11 g/mol
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00277447
CAS Number: 80517-21-1