2-Bromo-2'-(trifluoromethoxy)acetophenone 97%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(Trifluoromethoxy)phenacyl bromide
PC7361-1G PC7361-5G PC7361-25G PC7361-100G
APOSPC7361-1GEA 94.4 EUR
APOSPC7361-1G APOSPC7361-5G APOSPC7361-25G APOSPC7361-100G
2-Bromo-2'-(trifluoromethoxy)acetophenone 97%
2-Bromo-2'-(trifluoromethoxy)acetophenone
Formula: C₉H₆BrF₃O₂
Boiling Pt: 52 °C/0.04mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD03094424
CAS Number: 530141-40-3
UN: 3265
ADR: 8 III