2,2,2-trifluoroethylhydrazine 70% in aqueous solution
Supplier: Apollo Scientific
Warning
Synonyms: (2,2,2-Trifluoroethyl)hydrazine, 2,2,2-Trifluoroethylhydrazine
PC7366-1G PC7366-250MG PC7366-5G
APOSPC7366-1GEA 26.4 EUR
APOSPC7366-1G APOSPC7366-250MG APOSPC7366-5G
2,2,2-trifluoroethylhydrazine 70% in aqueous solution
2,2,2-trifluoroethylhydrazine
Formula: C₂H₅F₃N₂
Density: 1.294 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00007622
CAS Number: 5042-30-8
EINECS: 225-734-0
UN: 1993
ADR: 3,III