2,2,2-Trifluoroethyl trichloromethanesulphonate 97%
Supplier: Apollo Scientific
Danger
PC7377-1G PC7377-5G
APOSPC7377-1GEA 50.9 EUR
APOSPC7377-1G APOSPC7377-5G
2,2,2-Trifluoroethyl trichloromethanesulphonate 97%
2,2,2-Trifluoroethyl trichloromethanesulphonate
Formula: C₃H₂Cl₃F₃O₃S
MW: 281.47 g/mol
Boiling Pt: 84…86 °C/20mmHg
Melting Pt: –70 °C
Density: 1.714 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042400
CAS Number: 23199-56-6
EINECS: 245-484-6
UN: 2927
ADR: 6.1+8,II